Product Details

SNP ID
rs145811554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:45984326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCAGGCGCGCTGCACTACAGTGA[C/T]GAGGTGGAGATCATCCAAGGCCTCA
Phenotype
MIM: 120220
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL6A1 PubMed Links

Gene Details

Gene
COL6A1
Gene Name
collagen type VI alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001848.2 387 Silent Mutation GAC,GAT D95D NP_001839.2
Gene
LOC101928796
Gene Name
uncharacterized LOC101928796
There are no transcripts associated with this gene.

View Full Product Details