Product Details

SNP ID
rs146110723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41798989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTCTTTAGAGTTCTCACTTAT[A/G]ACAAGAAGTAAGAGGTGTCTGTGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRDM15 PubMed Links

Gene Details

Gene
PRDM15
Gene Name
PR domain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040424.2 5959 UTR 3 NP_001035514.1
NM_001282934.1 5959 UTR 3 NP_001269863.1
NM_022115.4 5959 Intron NP_071398.3
XM_006724039.2 5959 UTR 3 XP_006724102.1
XM_006724040.2 5959 UTR 3 XP_006724103.1
XM_011529674.1 5959 UTR 3 XP_011527976.1
XM_011529675.1 5959 UTR 3 XP_011527977.1
XM_011529676.2 5959 UTR 3 XP_011527978.1
XM_011529677.2 5959 UTR 3 XP_011527979.1
XM_011529678.2 5959 UTR 3 XP_011527980.1
XM_011529679.2 5959 UTR 3 XP_011527981.1
XM_011529681.2 5959 UTR 3 XP_011527983.1
XM_011529683.1 5959 UTR 3 XP_011527985.1
XM_017028425.1 5959 UTR 3 XP_016883914.1
XM_017028426.1 5959 UTR 3 XP_016883915.1

View Full Product Details