Product Details

SNP ID
rs150268877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:17790903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTGTCTTATCTCACTCCCCTT[A/C]ATCATATAAGCTCTGAAAATTTAGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C21orf91 PubMed Links

Gene Details

Gene
C21orf91
Gene Name
chromosome 21 open reading frame 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100420.1 3494 UTR 3 NP_001093890.1
NM_001100421.1 3494 UTR 3 NP_001093891.1
NM_017447.3 3494 UTR 3 NP_059143.3
Gene
C21orf91-OT1
Gene Name
C21orf91 overlapping transcript 1
There are no transcripts associated with this gene.

View Full Product Details