Product Details

SNP ID
rs144405404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21671773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACAGATGTATCAACATTTATGA[C/T]GGAGTGGTGGCTTATGTCTGTAGTC
Phenotype
MIM: 607588
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107985532 PubMed Links

Gene Details

Gene
LOC107985532
Gene Name
uncharacterized LOC107985532
There are no transcripts associated with this gene.

Gene
PPIL2
Gene Name
peptidylprolyl isomerase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317996.1 Intron NP_001304925.1
NM_014337.3 Intron NP_055152.1
NM_148175.2 Intron NP_680480.1
NM_148176.2 Intron NP_680481.1
XM_005261448.3 Intron XP_005261505.1
XM_011530041.2 Intron XP_011528343.1
XM_011530042.2 Intron XP_011528344.1
XM_011530043.2 Intron XP_011528345.1
XM_011530044.2 Intron XP_011528346.1
XM_011530046.2 Intron XP_011528348.1
XM_011530047.2 Intron XP_011528349.1
XM_011530049.2 Intron XP_011528351.1
XM_011530050.2 Intron XP_011528352.1
XM_011530051.2 Intron XP_011528353.1
XM_017028705.1 Intron XP_016884194.1
XM_017028706.1 Intron XP_016884195.1

View Full Product Details