Product Details

SNP ID
rs145756479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50739370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGGCCTGGCCCTGGATGGTCAG[C/T]CTCCAGATCTTCACGTACAACAGCC
Phenotype
MIM: 102480 MIM: 606230
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACR PubMed Links

Gene Details

Gene
ACR
Gene Name
acrosin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097.2 189 Silent Mutation AGC,AGT S59S NP_001088.2
Gene
LOC105373100
Gene Name
uncharacterized LOC105373100
There are no transcripts associated with this gene.

Gene
SHANK3
Gene Name
SH3 and multiple ankyrin repeat domains 3
There are no transcripts associated with this gene.

View Full Product Details