Product Details

SNP ID
rs145956190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43881630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCATGTTCCTCAGCAGGCCCTGC[A/G]TCCACCACAAGTCCGCCGGCACATC
Phenotype
MIM: 611589
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNPLA5 PubMed Links

Gene Details

Gene
PNPLA5
Gene Name
patatin like phospholipase domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177675.1 918 Missense Mutation NP_001171146.1
NM_138814.3 918 Missense Mutation NP_620169.1
XM_011529959.2 918 Missense Mutation XP_011528261.1

View Full Product Details