Product Details

SNP ID
rs147251834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23181596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCAGGCCATGCAGATGGAGCG[C/T]AAAAAGTCCCAGCACGGCGCGGGCT
Phenotype
MIM: 151410 MIM: 605663
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCR PubMed Links

Gene Details

Gene
BCR
Gene Name
BCR, RhoGEF and GTPase activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004327.3 1232 Silent Mutation CGC,CGT R212R NP_004318.3
NM_021574.2 1232 Silent Mutation CGC,CGT R212R NP_067585.2
Gene
RSPH14
Gene Name
radial spoke head 14 homolog
There are no transcripts associated with this gene.

View Full Product Details