Product Details

SNP ID
rs138065939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112909553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACTACACTAGGTAGCACCCCCT[A/G]TTAAAGACAAACTCTTCTCTCTCAT
Phenotype
MIM: 605510
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL37 PubMed Links

Gene Details

Gene
IL37
Gene Name
interleukin 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014439.3 665 Intron NP_055254.2
NM_173202.1 665 Intron NP_775294.1
NM_173203.1 665 Intron NP_775295.1
NM_173204.1 665 Intron NP_775296.1
NM_173205.1 665 Intron NP_775297.1
XM_011510963.2 665 UTR 5 XP_011509265.1
XM_011510964.2 665 UTR 5 XP_011509266.1

View Full Product Details