Product Details

SNP ID
rs140253994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127183940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCCTGCTTCCACAGTTAACAAC[A/T]TCTGCCTAATCTTGGGTCATCTCTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP27C1 PubMed Links

Gene Details

Gene
CYP27C1
Gene Name
cytochrome P450 family 27 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001665.3 4581 UTR 3 NP_001001665.3
XM_017003960.1 4581 UTR 3 XP_016859449.1
XM_017003961.1 4581 Intron XP_016859450.1

View Full Product Details