Product Details

SNP ID
rs141225794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74474648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGCCTGCTGATTTCCCACCAG[A/G]TAGCCCTCCGGGCTAGGTCCCCTTC
Phenotype
MIM: 601336 MIM: 611857
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC142 PubMed Links

Gene Details

Gene
CCDC142
Gene Name
coiled-coil domain containing 142
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032779.3 2523 Silent Mutation TAC,TAT Y710Y NP_116168.3
Gene
MOGS
Gene Name
mannosyl-oligosaccharide glucosidase
There are no transcripts associated with this gene.

Gene
MRPL53
Gene Name
mitochondrial ribosomal protein L53
There are no transcripts associated with this gene.

Gene
TTC31
Gene Name
tetratricopeptide repeat domain 31
There are no transcripts associated with this gene.

View Full Product Details