Product Details

SNP ID
rs141716313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202377541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATGGACCATCCTGCTGGTCAGC[A/G]CTGCGGCTGGTGAGTAGCTCCGGCC
Phenotype
MIM: 600799
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMPR2 PubMed Links

Gene Details

Gene
BMPR2
Gene Name
bone morphogenetic protein receptor type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204.6 1215 Missense Mutation ACT,GCT T23A NP_001195.2
XM_011511687.1 1215 Missense Mutation ACT,GCT T23A XP_011509989.1

View Full Product Details