Product Details

SNP ID
rs142143860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219077324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCTGGTTTACACATTGGCTATC[A/G]ATTCCTTGCAGGGAAGCACTGTTTG
Phenotype
MIM: 611290
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NHEJ1 PubMed Links

Gene Details

Gene
NHEJ1
Gene Name
non-homologous end joining factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024782.2 893 Silent Mutation NP_079058.1

View Full Product Details