Product Details

SNP ID
rs143080277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:105749599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACCAACGTGTTGCTGGGTAGAG[C/T]TGTTTTTTTGTGAGGCAGTTTACAG
Phenotype
MIM: 604930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCK2 PubMed Links

Gene Details

Gene
NCK2
Gene Name
NCK adaptor protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004720.2 Intron NP_001004720.1
NM_001004722.3 Intron NP_001004722.1
NM_003581.4 Intron NP_003572.2
XM_006712797.3 Intron XP_006712860.1
XM_011511991.2 Intron XP_011510293.1
XM_011511992.2 Intron XP_011510294.2
XM_017005103.1 Intron XP_016860592.1
XM_017005104.1 Intron XP_016860593.1
XM_017005105.1 Intron XP_016860594.1

View Full Product Details