Product Details

SNP ID
rs144183463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:19994768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCCAGGGATATAGAAATAAGAC[A/C]ATGTTTATGTAATACTGTGTAATTA
Phenotype
MIM: 602109 MIM: 613602
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928222 PubMed Links

Gene Details

Gene
LOC101928222
Gene Name
uncharacterized LOC101928222
There are no transcripts associated with this gene.

Gene
MATN3
Gene Name
matrilin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002381.4 Intron NP_002372.1
Gene
WDR35
Gene Name
WD repeat domain 35
There are no transcripts associated with this gene.

View Full Product Details