Product Details

SNP ID
rs144543979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96266481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGACCGGAGAATCCGCATCTGGG[C/G]CACGGAGGGTAAGGCCCAGCCTGGT
Phenotype
MIM: 604333 MIM: 601664 MIM: 613403
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CIAO1 PubMed Links

Gene Details

Gene
CIAO1
Gene Name
cytosolic iron-sulfur assembly component 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004804.2 336 Missense Mutation GCC,GGC A44G NP_004795.1
Gene
SNRNP200
Gene Name
small nuclear ribonucleoprotein U5 subunit 200
There are no transcripts associated with this gene.

Gene
TMEM127
Gene Name
transmembrane protein 127
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193304.2 336 Intron NP_001180233.1
NM_017849.3 336 Intron NP_060319.1
XM_017004450.1 336 Intron XP_016859939.1
XM_017004451.1 336 Intron XP_016859940.1
XM_017004452.1 336 Intron XP_016859941.1

View Full Product Details