Product Details

SNP ID
rs147026593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70979271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCTACTACCAGCTGTTCGCAGC[A/G]GCTGTGGGCAACGTGGAATGGCTGC
Phenotype
MIM: 617009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD53 PubMed Links

Gene Details

Gene
ANKRD53
Gene Name
ankyrin repeat domain 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001115116.1 382 Silent Mutation GCA,GCG A115A NP_001108588.1
NM_024933.3 382 Silent Mutation GCA,GCG A115A NP_079209.3
XM_005264569.4 382 Silent Mutation GCA,GCG A81A XP_005264626.1
XM_011533114.2 382 Silent Mutation GCA,GCG A115A XP_011531416.1
Gene
TEX261
Gene Name
testis expressed 261
There are no transcripts associated with this gene.

View Full Product Details