Product Details

SNP ID
rs148252568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219075665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCAGAAGGATGAGGCGGGAGGA[C/T]TGATAAACTACCCCACGTGGTAGTT
Phenotype
MIM: 611290
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NHEJ1 PubMed Links

Gene Details

Gene
NHEJ1
Gene Name
non-homologous end joining factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024782.2 1762 UTR 3 NP_079058.1

View Full Product Details