Product Details

SNP ID
rs148816283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99018285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACATTTTGTATTTGGCTTCATTG[G/T]AGAACTCATATGATAAGCATTACTT
Phenotype
MIM: 607166
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TSGA10 PubMed Links

Gene Details

Gene
TSGA10
Gene Name
testis specific 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025244.2 2369 Missense Mutation NP_079520.1
NM_182911.3 2369 Missense Mutation NP_878915.2
XM_005264023.1 2369 Missense Mutation XP_005264080.1
XM_005264024.2 2369 Missense Mutation XP_005264081.1
XM_005264025.1 2369 Missense Mutation XP_005264082.1
XM_005264026.1 2369 Missense Mutation XP_005264083.1
XM_005264027.1 2369 Missense Mutation XP_005264084.1
XM_005264029.1 2369 Missense Mutation XP_005264086.1
XM_006712781.2 2369 Missense Mutation XP_006712844.1
XM_011511934.1 2369 Missense Mutation XP_011510236.1
XM_011511935.2 2369 Missense Mutation XP_011510237.1
XM_011511937.1 2369 Missense Mutation XP_011510239.1
XM_011511939.1 2369 Missense Mutation XP_011510241.1
XM_011511940.2 2369 Missense Mutation XP_011510242.1
XM_011511941.1 2369 Missense Mutation XP_011510243.1
XM_011511942.1 2369 Missense Mutation XP_011510244.1
XM_011511943.2 2369 Missense Mutation XP_011510245.1
XM_017005035.1 2369 Missense Mutation XP_016860524.1
XM_017005036.1 2369 Intron XP_016860525.1
XM_017005037.1 2369 Intron XP_016860526.1
XM_017005038.1 2369 Intron XP_016860527.1
XM_017005039.1 2369 Missense Mutation XP_016860528.1
XM_017005040.1 2369 Missense Mutation XP_016860529.1
XM_017005041.1 2369 Missense Mutation XP_016860530.1
XM_017005042.1 2369 Missense Mutation XP_016860531.1
XM_017005043.1 2369 Missense Mutation XP_016860532.1
XM_017005044.1 2369 Missense Mutation XP_016860533.1
XM_017005045.1 2369 Missense Mutation XP_016860534.1
XM_017005046.1 2369 Missense Mutation XP_016860535.1
XM_017005047.1 2369 Missense Mutation XP_016860536.1
XM_017005048.1 2369 Missense Mutation XP_016860537.1

View Full Product Details