Product Details

SNP ID
rs138135409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10249209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCTTGCAGGTGCCCATGGCGTC[C/T]GAGCGGGGCAAGGTCAAGCACAACT
Phenotype
MIM: 603304
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRAK2 PubMed Links

Gene Details

Gene
IRAK2
Gene Name
interleukin 1 receptor associated kinase 2
There are no transcripts associated with this gene.

Gene
TATDN2
Gene Name
TatD DNase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014760.3 590 Silent Mutation TCC,TCT S3S NP_055575.3

View Full Product Details