Product Details

SNP ID
rs138535647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112141024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGGCTTTAAGACTAAAATTTAC[C/T]CTAAGATTTTCTAAAATGTTTTGTA
Phenotype
MIM: 611956 MIM: 607792 MIM: 612738
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C3orf52 PubMed Links

Gene Details

Gene
C3orf52
Gene Name
chromosome 3 open reading frame 52
There are no transcripts associated with this gene.

Gene
GCSAM
Gene Name
germinal center associated signaling and motility
There are no transcripts associated with this gene.

Gene
SLC9C1
Gene Name
solute carrier family 9 member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320531.1 3849 UTR 3 NP_001307460.1
NM_183061.2 3849 UTR 3 NP_898884.1
XM_006713603.2 3849 UTR 3 XP_006713666.1
XM_011512718.1 3849 UTR 3 XP_011511020.1
XM_011512719.1 3849 UTR 3 XP_011511021.1
XM_011512720.1 3849 UTR 3 XP_011511022.1
XM_011512721.1 3849 UTR 3 XP_011511023.1
XM_011512724.1 3849 Intron XP_011511026.1
XM_011512725.2 3849 UTR 3 XP_011511027.1
XM_011512726.2 3849 Intron XP_011511028.1
XM_011512727.2 3849 Intron XP_011511029.1
XM_017006246.1 3849 UTR 3 XP_016861735.1
XM_017006247.1 3849 UTR 3 XP_016861736.1
XM_017006248.1 3849 UTR 3 XP_016861737.1
XM_017006249.1 3849 Intron XP_016861738.1
XM_017006250.1 3849 Intron XP_016861739.1

View Full Product Details