Product Details

SNP ID
rs139342438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195748975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTAGCCCCTACTGCACGGGGACA[C/T]GCAGGTGAAGCCGCTCTGGGGGCTG
Phenotype
MIM: 158372
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MUC4 PubMed Links

Gene Details

Gene
MUC4
Gene Name
mucin 4, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322468.1 22135 Missense Mutation NP_001309397.1
NM_004532.5 22135 Missense Mutation ATG,GTG M1085V NP_004523.3
NM_018406.6 22135 Intron NP_060876.5
NM_138297.4 22135 Missense Mutation ATG,GTG M1034V NP_612154.2

View Full Product Details