Product Details

SNP ID
rs139500325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49109191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCCAAATCCGGGAAGCCTAGG[A/G]TATGTGGAAACGGCCATCAGCACCC
Phenotype
MIM: 603727 MIM: 614471
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6890 PubMed Links

Gene Details

Gene
MIR6890
Gene Name
microRNA 6890
There are no transcripts associated with this gene.

Gene
QARS
Gene Name
glutaminyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
USP19
Gene Name
ubiquitin specific peptidase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199160.1 Intron NP_001186089.1
NM_001199161.1 Intron NP_001186090.1
NM_001199162.1 Intron NP_001186091.1
NM_006677.2 Intron NP_006668.1
XM_005264823.2 Intron XP_005264880.1
XM_005264824.2 Intron XP_005264881.1
XM_005264825.2 Intron XP_005264882.1
XM_005264826.2 Intron XP_005264883.1
XM_005264827.2 Intron XP_005264884.1
XM_005264829.2 Intron XP_005264886.1
XM_005264830.2 Intron XP_005264887.1
XM_005264831.2 Intron XP_005264888.1
XM_006712946.2 Intron XP_006713009.1
XM_006712947.2 Intron XP_006713010.1
XM_006712948.2 Intron XP_006713011.1
XM_006712949.2 Intron XP_006713012.1
XM_006712950.2 Intron XP_006713013.1
XM_006712951.2 Intron XP_006713014.1
XM_006712952.2 Intron XP_006713015.1
XM_006712953.1 Intron XP_006713016.1
XM_017005615.1 Intron XP_016861104.1
XM_017005616.1 Intron XP_016861105.1
XM_017005617.1 Intron XP_016861106.1
XM_017005618.1 Intron XP_016861107.1
XM_017005619.1 Intron XP_016861108.1
XM_017005620.1 Intron XP_016861109.1
XM_017005621.1 Intron XP_016861110.1
XM_017005622.1 Intron XP_016861111.1
XM_017005623.1 Intron XP_016861112.1
XM_017005624.1 Intron XP_016861113.1
XM_017005625.1 Intron XP_016861114.1
XM_017005626.1 Intron XP_016861115.1
XM_017005627.1 Intron XP_016861116.1
XM_017005628.1 Intron XP_016861117.1
XM_017005629.1 Intron XP_016861118.1
XM_017005630.1 Intron XP_016861119.1
XM_017005631.1 Intron XP_016861120.1
XM_017005632.1 Intron XP_016861121.1
XM_017005633.1 Intron XP_016861122.1
XM_017005634.1 Intron XP_016861123.1
XM_017005635.1 Intron XP_016861124.1

View Full Product Details