Product Details

SNP ID
rs139533837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44718515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAAGTCCTTAAAATGTTTTTTAG[A/G]GTTCTGTTTGAACTGGCTTGCCTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF502 PubMed Links

Gene Details

Gene
ZNF502
Gene Name
zinc finger protein 502
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134440.1 Intron NP_001127912.1
NM_001134441.1 Intron NP_001127913.1
NM_001134442.1 Intron NP_001127914.1
NM_001282880.1 Intron NP_001269809.1
NM_033210.4 Intron NP_149987.2
XM_017007440.1 Intron XP_016862929.1

View Full Product Details