Product Details

SNP ID
rs145449375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195747255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCAGAAGCGCAGGACCACGAAC[G/T]TCCCGACCCCCAGCAGCAAGAGGCC
Phenotype
MIM: 158372
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MUC4 PubMed Links

Gene Details

Gene
MUC4
Gene Name
mucin 4, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322468.1 22334 Missense Mutation AGG,ATG R7393M NP_001309397.1
NM_004532.5 22334 Missense Mutation AGG,ATG R1151M NP_004523.3
NM_018406.6 22334 Intron AGG,ATG R5387M NP_060876.5
NM_138297.4 22334 Missense Mutation AGG,ATG R1100M NP_612154.2

View Full Product Details