Product Details
- SNP ID
-
rs146246326
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:195571307 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAACTTAACTTCCAGCTTGGCAG[C/G]CTCTGTGAGGTTAACTGGGGTGGCT
- Phenotype
-
MIM: 107740
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
APOD
PubMed Links
Gene Details
- Gene
- APOD
- Gene Name
- apolipoprotein D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001647.3 |
666 |
Missense Mutation |
CCT,GCT |
P102A |
NP_001638.1 |
View Full Product Details