Product Details

SNP ID
rs146541622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44721118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACCCCCTGAAATTATTAGTCAA[G/T]GATATAATTTTGAGAAAAGCTTGCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF501 PubMed Links

Gene Details

Gene
ZNF501
Gene Name
zinc finger protein 501
There are no transcripts associated with this gene.

Gene
ZNF502
Gene Name
zinc finger protein 502
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134440.1 1798 Nonsense Mutation GGA,TGA G101* NP_001127912.1
NM_001134441.1 1798 Nonsense Mutation GGA,TGA G101* NP_001127913.1
NM_001134442.1 1798 Nonsense Mutation GGA,TGA G101* NP_001127914.1
NM_001282880.1 1798 Nonsense Mutation GGA,TGA G101* NP_001269809.1
NM_033210.4 1798 Nonsense Mutation GGA,TGA G101* NP_149987.2
XM_017007440.1 1798 Nonsense Mutation GGA,TGA G101* XP_016862929.1

View Full Product Details