Product Details

SNP ID
rs147382619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151814287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGATAAACGCACATCTGAAAA[C/G]TATACAAAATTTGGTAAGTTTGGAA
Phenotype
MIM: 600338
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AADAC PubMed Links

Gene Details

Gene
AADAC
Gene Name
arylacetamide deacetylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001086.2 215 Missense Mutation ACT,AGT T42S NP_001077.2
XM_005247104.4 215 Missense Mutation ACT,AGT T42S XP_005247161.1
Gene
AADACL2-AS1
Gene Name
AADACL2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details