Product Details

SNP ID
rs147486673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10249262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCACGTCGGAAGGGTGTCCCC[A/G]CAAGCGCAGCTGCCTCCGGGAGCCC
Phenotype
MIM: 603304
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRAK2 PubMed Links

Gene Details

Gene
IRAK2
Gene Name
interleukin 1 receptor associated kinase 2
There are no transcripts associated with this gene.

Gene
TATDN2
Gene Name
TatD DNase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014760.3 643 Missense Mutation CAC,CGC H21R NP_055575.3

View Full Product Details