Product Details

SNP ID
rs148896847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38123094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGCAGCGGCTCCCATTCCAGTCC[C/T]GATGCACATGGACACCACTCCGTAT
Phenotype
MIM: 604054 MIM: 604050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAA1 PubMed Links

Gene Details

Gene
ACAA1
Gene Name
acetyl-CoA acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130410.1 1435 Missense Mutation AGG,GGG R317G NP_001123882.1
NM_001607.3 1435 Missense Mutation AGG,GGG R410G NP_001598.1
XM_006713122.1 1435 Missense Mutation AGG,GGG R350G XP_006713185.1
XM_006713123.2 1435 Missense Mutation AGG,GGG R258G XP_006713186.1
XM_011533650.2 1435 Missense Mutation AGG,GGG R265G XP_011531952.1
Gene
DLEC1
Gene Name
deleted in lung and esophageal cancer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321153.1 1435 Intron NP_001308082.1
NM_007335.3 1435 Intron NP_031361.2
NM_007337.3 1435 Intron NP_031363.2
XM_006713438.3 1435 Intron XP_006713501.1
XM_006713439.3 1435 Intron XP_006713502.1
XM_006713440.3 1435 Intron XP_006713503.1
XM_011534317.2 1435 Intron XP_011532619.1
XM_011534318.2 1435 Intron XP_011532620.1

View Full Product Details