Product Details

SNP ID
rs150134287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119146443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAAAATGGGCATAGTACACCTG[A/G]TCAGGCTGGCCGCAGAGCATCCAAC
Phenotype
MIM: 608351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf30 PubMed Links

Gene Details

Gene
C3orf30
Gene Name
chromosome 3 open reading frame 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152539.2 294 Missense Mutation GAT,GGT D85G NP_689752.2
Gene
IGSF11
Gene Name
immunoglobulin superfamily member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015887.1 294 Intron NP_001015887.1
NM_152538.2 294 Intron NP_689751.2
XM_006713517.1 294 Intron XP_006713580.1
XM_011512462.2 294 Intron XP_011510764.1
XM_011512463.1 294 Intron XP_011510765.1
XM_011512464.2 294 Intron XP_011510766.1
XM_011512465.1 294 Intron XP_011510767.1
XM_011512466.2 294 Intron XP_011510768.1
XM_011512467.2 294 Intron XP_011510769.1
XM_011512468.1 294 Intron XP_011510770.1
XM_011512469.1 294 Intron XP_011510771.1
XM_011512470.1 294 Intron XP_011510772.1
XM_011512471.2 294 Intron XP_011510773.1
XM_011512472.1 294 Intron XP_011510774.1
XM_011512474.2 294 Intron XP_011510776.1
XM_017005789.1 294 Intron XP_016861278.1
XM_017005790.1 294 Intron XP_016861279.1

View Full Product Details