Product Details

SNP ID
rs138386005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:372823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAGTGTAAGTTGCAGAAAGGAG[A/G]TTATAATGAATTTAATGAATGCTTG
Phenotype
MIM: 194648
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF141 PubMed Links

Gene Details

Gene
ZNF141
Gene Name
zinc finger protein 141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003441.2 1152 Missense Mutation GAT,GGT D129G NP_003432.1
XM_011513562.2 1152 Missense Mutation GAT,GGT D157G XP_011511864.1
XM_011513563.2 1152 Missense Mutation GAT,GGT D53G XP_011511865.1
XM_017008591.1 1152 Missense Mutation GAT,GGT D189G XP_016864080.1

View Full Product Details