Product Details

SNP ID
rs139688335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15936293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCGCAGATTCCGGGCAACTTGTT[C/T]CCAATAGACTCTCTCATCCTTGAGC
Phenotype
MIM: 607737
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGFBP1 PubMed Links

Gene Details

Gene
FGFBP1
Gene Name
fibroblast growth factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005130.4 635 Missense Mutation AAA,GAA K114E NP_005121.1

View Full Product Details