Product Details

SNP ID
rs139936568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15936050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATCTGGGTCCTCCACACACTCGG[G/T]AGCTTTGGTGGCCATGGTCTGGGTC
Phenotype
MIM: 607737
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FGFBP1 PubMed Links

Gene Details

Gene
FGFBP1
Gene Name
fibroblast growth factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005130.4 878 Missense Mutation ACC,CCC T195P NP_005121.1

View Full Product Details