Product Details

SNP ID
rs140348838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17813092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAATGTGAGAATACAGGCAGTTC[C/T]GGCGCTTTCACGACTTCAGGATCCC
Phenotype
MIM: 606280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAF16 PubMed Links

Gene Details

Gene
DCAF16
Gene Name
DDB1 and CUL4 associated factor 16
There are no transcripts associated with this gene.

Gene
NCAPG
Gene Name
non-SMC condensin I complex subunit G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022346.4 756 Missense Mutation CCG,CTG P164L NP_071741.2
XM_011513876.2 756 UTR 5 XP_011512178.1
XM_017008543.1 756 Missense Mutation CCG,CTG P164L XP_016864032.1
XM_017008544.1 756 UTR 5 XP_016864033.1

View Full Product Details