Product Details

SNP ID
rs141176394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:110618461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTGGGGAAAACATGCTCTGTGA[A/T]GACAGGGGGTTGACGTTCATAGAGT
Phenotype
MIM: 601542
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PITX2 PubMed Links

Gene Details

Gene
PITX2
Gene Name
paired like homeodomain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000325.5 2069 Silent Mutation TCA,TCT S213S NP_000316.2
NM_001204397.1 2069 Silent Mutation TCA,TCT S206S NP_001191326.1
NM_001204398.1 2069 Silent Mutation TCA,TCT S206S NP_001191327.1
NM_001204399.1 2069 Silent Mutation TCA,TCT S160S NP_001191328.1
NM_153426.2 2069 Silent Mutation TCA,TCT S206S NP_700475.1
NM_153427.2 2069 Silent Mutation TCA,TCT S160S NP_700476.1
XM_011532027.2 2069 Silent Mutation TCA,TCT S160S XP_011530329.1

View Full Product Details