Product Details

SNP ID
rs142232040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:168092708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAACAATTACCATCAAAATGTTTT[G/T]TGGAGACTATGTGAGTATAATGCTT
Phenotype
MIM: 608008
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANXA10 PubMed Links

Gene Details

Gene
ANXA10
Gene Name
annexin A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007193.4 172 Missense Mutation TGT,TTT C3F NP_009124.2
XM_011531571.2 172 Missense Mutation TGT,TTT C3F XP_011529873.1

View Full Product Details