Product Details
- SNP ID
-
rs146370385
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:3766918 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGACATCCTGGTGGCCACGCTGGT[A/C]ATGCCCTTCTCGTTGGCCAACGAGC
- Phenotype
-
MIM: 104250
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADRA2C
PubMed Links
Gene Details
- Gene
- ADRA2C
- Gene Name
- adrenoceptor alpha 2C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000683.3 |
350 |
Silent Mutation |
GTA,GTC |
V104V |
NP_000674.2 |
View Full Product Details