Product Details

SNP ID
rs146859271
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15936094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGTCACTGCTAGGCTAGAGGGG[C/G]TGGTCTCTTTGCCTTTGATGTGCTC
Phenotype
MIM: 607737
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
FGFBP1 PubMed Links

Gene Details

Gene
FGFBP1
Gene Name
fibroblast growth factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005130.4 834 Missense Mutation ACC,AGC T180S NP_005121.1

View Full Product Details