Product Details
- SNP ID
-
rs147527227
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:56338763 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTGAGGTACACGGGGATGAAAAG[A/G]TTGGTCCACTGGTAGAGAACTGCCA
- Phenotype
-
MIM: 614365
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AASDH
PubMed Links
Gene Details
- Gene
- AASDH
- Gene Name
- aminoadipate-semialdehyde dehydrogenase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286668.1 |
2901 |
Missense Mutation |
ACC,ATC |
T879I |
NP_001273597.1 |
NM_001286669.1 |
2901 |
Missense Mutation |
ACC,ATC |
T826I |
NP_001273598.1 |
NM_001286670.1 |
2901 |
Missense Mutation |
ACC,ATC |
T494I |
NP_001273599.1 |
NM_001286671.1 |
2901 |
Silent Mutation |
AAC,AAT |
N839N |
NP_001273600.1 |
NM_001286672.1 |
2901 |
Intron |
|
|
NP_001273601.1 |
NM_001323890.1 |
2901 |
Missense Mutation |
ACC,ATC |
T1018I |
NP_001310819.1 |
NM_001323892.1 |
2901 |
Silent Mutation |
AAC,AAT |
N924N |
NP_001310821.1 |
NM_001323893.1 |
2901 |
Silent Mutation |
AAC,AAT |
N771N |
NP_001310822.1 |
NM_001323899.1 |
2901 |
Missense Mutation |
ACC,ATC |
T602I |
NP_001310828.1 |
NM_181806.3 |
2901 |
Missense Mutation |
ACC,ATC |
T979I |
NP_861522.2 |
XM_017007740.1 |
2901 |
Missense Mutation |
ACC,ATC |
T935I |
XP_016863229.1 |
XM_017007741.1 |
2901 |
Missense Mutation |
ACC,ATC |
T826I |
XP_016863230.1 |
XM_017007742.1 |
2901 |
Missense Mutation |
ACC,ATC |
T826I |
XP_016863231.1 |
XM_017007743.1 |
2901 |
Silent Mutation |
AAC,AAT |
N880N |
XP_016863232.1 |
XM_017007744.1 |
2901 |
Missense Mutation |
ACC,ATC |
T557I |
XP_016863233.1 |
- Gene
- KIAA1211
- Gene Name
- KIAA1211
There are no transcripts associated with this gene.
View Full Product Details