Product Details

SNP ID
rs147869831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17803392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATTGGCTTGCCAGGGCATAAGA[C/G]AGTTTGACTGAGAAGGCATTAGTGG
Phenotype
MIM: 606280
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCAF16 PubMed Links

Gene Details

Gene
DCAF16
Gene Name
DDB1 and CUL4 associated factor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017741.3 1695 UTR 3 NP_060211.3
XM_005248169.3 1695 UTR 3 XP_005248226.1
XM_005248171.3 1695 UTR 3 XP_005248228.1
XM_006713966.3 1695 UTR 3 XP_006714029.1
XM_017008327.1 1695 UTR 3 XP_016863816.1
XM_017008328.1 1695 UTR 3 XP_016863817.1
Gene
NCAPG
Gene Name
non-SMC condensin I complex subunit G
There are no transcripts associated with this gene.

View Full Product Details