Product Details

SNP ID
rs148361115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48138712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCATACACATAGTTGGACGCCA[A/G]CTTCGGCTGGTAGAGTCGGTGGCCT
Phenotype
MIM: 600583 MIM: 600058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEC PubMed Links

Gene Details

Gene
TEC
Gene Name
tec protein tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003215.2 3062 Silent Mutation CTG,TTG L589L NP_003206.2
XM_011513737.2 3062 Silent Mutation CTG,TTG L553L XP_011512039.1
XM_011513741.1 3062 Intron XP_011512043.1
XM_017008569.1 3062 Silent Mutation CTG,TTG L473L XP_016864058.1
Gene
TXK
Gene Name
TXK tyrosine kinase
There are no transcripts associated with this gene.

View Full Product Details