Product Details

SNP ID
rs149422615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:74038151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGGGATGCGGGGTTGAGACAAG[A/C]TTTCTTCCCATTCTTGAGTGTGGCT
Phenotype
MIM: 139111
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CXCL3 PubMed Links

Gene Details

Gene
CXCL3
Gene Name
C-X-C motif chemokine ligand 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002090.2 412 Missense Mutation GCT,TCT A84S NP_002081.2

View Full Product Details