Product Details

SNP ID
rs149730292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:173388312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGCAGTTTGTGGGAAATCAGGA[A/G]TGGTGTTCTCCAGAATACATGAAGA
Phenotype
MIM: 603163
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCRG1 PubMed Links

Gene Details

Gene
SCRG1
Gene Name
stimulator of chondrogenesis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011531578.1 452 UTR 3 XP_011529880.1
XM_017007678.1 452 UTR 3 XP_016863167.1

View Full Product Details