Product Details

SNP ID
rs149848389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48892851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTACTCTGGCATACTCCTATGT[A/G]TGATACCTTTCCAAGGCCAAATCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OCIAD2 PubMed Links

Gene Details

Gene
OCIAD2
Gene Name
OCIA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014446.2 537 Missense Mutation CAC,TAC H102Y NP_001014446.1
NM_001286773.1 537 Intron NP_001273702.1
NM_001286774.1 537 Missense Mutation CAC,TAC H40Y NP_001273703.1
NM_152398.3 537 Intron NP_689611.1

View Full Product Details