Product Details

SNP ID
rs150580368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70688930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGAGGAGGAGGAGGTGCTAGCC[C/T]TAGATATGGACGATGAGGACGACGA
Phenotype
MIM: 611614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UTP3 PubMed Links

Gene Details

Gene
UTP3
Gene Name
UTP3, small subunit processome component homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020368.2 452 Silent Mutation CTA,TTA L85L NP_065101.1

View Full Product Details