Product Details

SNP ID
rs142783315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179798348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTCTCCCCCAAACCCTACCCACC[A/G]ATCTGGAGGTAGCCGTCGGGGCTCC
Phenotype
MIM: 246530 MIM: 604561 MIM: 601530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LTC4S PubMed Links

Gene Details

Gene
LTC4S
Gene Name
leukotriene C4 synthase
There are no transcripts associated with this gene.

Gene
MGAT4B
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014275.4 1704 Silent Mutation ATC,ATT I503I NP_055090.1
NM_054013.3 1704 Silent Mutation ATC,ATT I518I NP_463459.1
XM_017008998.1 1704 Silent Mutation ATC,ATT I503I XP_016864487.1
Gene
MIR1229
Gene Name
microRNA 1229
There are no transcripts associated with this gene.

Gene
SQSTM1
Gene Name
sequestosome 1
There are no transcripts associated with this gene.

View Full Product Details