Product Details

SNP ID
rs143788121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73557582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAAATAGGCTATTACCTTGTTC[A/G]ATACGAGTAGCCAGATAGAGCATCT
Phenotype
MIM: 605787 MIM: 616194
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRA2 PubMed Links

Gene Details

Gene
ANKRA2
Gene Name
ankyrin repeat family A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023039.4 1024 Silent Mutation ATC,ATT I169I NP_075526.1
XM_005248560.2 1024 Silent Mutation ATC,ATT I101I XP_005248617.1
XM_017009678.1 1024 Silent Mutation ATC,ATT I8I XP_016865167.1
Gene
UTP15
Gene Name
UTP15, small subunit processome component
There are no transcripts associated with this gene.

View Full Product Details