Product Details

SNP ID
rs145064087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95648425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTAGACTTAAAATCGAAACATA[G/T]TGGGAGTTTATGAATAGTTCATCAA
Phenotype
MIM: 608039
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RFESD PubMed Links

Gene Details

Gene
RFESD
Gene Name
Rieske Fe-S domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131065.1 Intron NP_001124537.1
NM_001131066.1 Intron NP_001124538.1
NM_173362.3 Intron NP_775498.1
XM_011543360.2 Intron XP_011541662.1
XM_011543361.2 Intron XP_011541663.1
XM_017009413.1 Intron XP_016864902.1
Gene
SPATA9
Gene Name
spermatogenesis associated 9
There are no transcripts associated with this gene.

View Full Product Details