Product Details

SNP ID
rs147028950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:57175024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCTCTTTACACTTATCTCTGA[A/G]TTACATTCGCTAATACTCCGTGGCA
Phenotype
MIM: 608412
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPBP1 PubMed Links

Gene Details

Gene
GPBP1
Gene Name
GC-rich promoter binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127235.2 Intron NP_001120707.2
NM_001127236.2 Intron NP_001120708.1
NM_001203246.1 Intron NP_001190175.1
NM_022913.3 Intron NP_075064.1
XM_006714673.3 Intron XP_006714736.1
XM_006714674.3 Intron XP_006714737.1
XM_017009754.1 Intron XP_016865243.1
XM_017009755.1 Intron XP_016865244.1
XM_017009756.1 Intron XP_016865245.1
XM_017009757.1 Intron XP_016865246.1
XM_017009758.1 Intron XP_016865247.1
XM_017009759.1 Intron XP_016865248.1

View Full Product Details