Product Details

SNP ID
rs148709770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:74769242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGATGCATGAAAAGAGAAACACC[A/G]AACAAAAGAATGATGAAAAGACACC
Phenotype
MIM: 615769 MIM: 606544 MIM: 612497
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM169A PubMed Links

Gene Details

Gene
FAM169A
Gene Name
family with sequence similarity 169 member A
There are no transcripts associated with this gene.

Gene
GFM2
Gene Name
G elongation factor mitochondrial 2
There are no transcripts associated with this gene.

Gene
NSA2
Gene Name
NSA2, ribosome biogenesis homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271665.1 590 Missense Mutation AAA,GAA K74E NP_001258594.1
NM_014886.4 590 Missense Mutation AAA,GAA K74E NP_055701.1
XM_011543098.1 590 Intron XP_011541400.1
XM_017008952.1 590 Intron XP_016864441.1
XM_017008953.1 590 Intron XP_016864442.1

View Full Product Details